siRNA Details
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013795.5 | NP_038823.2 | 2 | 253 | ||
GenBank mRNA | AK005175.1 | - | 2 | 200 | ||
AK005321.1 | - | 2 | 230 | |||
AK005380.1 | - | 2 | 231 | |||
AK008002.1 | - | 2 | 190 | |||
AK008012.1 | - | 2 | 227 | |||
AK008030.1 | - | 2 | 216 | |||
AK008036.1 | - | 2 | 230 | |||
AK008087.1 | - | 2 | 204 | |||
AK008169.1 | - | 2 | 230 | |||
AK008183.1 | - | 2 | 218 | |||
AK008251.1 | - | 2 | 218 | |||
AK008321.1 | - | 2 | 226 | |||
AK009465.1 | - | 2 | 219 | |||
AK009468.1 | - | 2 | 219 | |||
AK009471.1 | - | 2 | 219 | |||
AK009612.1 | - | 2 | 212 | |||
AK009806.1 | - | 2 | 218 | |||
AK011038.1 | - | 2 | 197 | |||
AK011046.1 | - | 2 | 197 | |||
AK011099.1 | - | 2 | 224 | |||
AK011842.1 | - | 2 | 190 | |||
AK019252.1 | - | 2 | 219 | |||
AK019274.1 | - | 2 | 219 | |||
AK019320.1 | - | 2 | 226 | |||
AK019343.1 | - | 2 | 190 | |||
AK019350.1 | - | 2 | 218 | |||
AK019355.1 | - | 2 | 218 | |||
AK146520.1 | - | 2323 | ||||
BC031384.1 | - | 2 | 226 | 5356491 | ||
BC086880.1 | - | 2 | 189 | 6762989 | ||
BC089555.1 | - | 2 | 229 | 30312783 | ||
BC094407.1 | - | 2 | 183 | 573475 | ||
BY269662.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
