siRNA Details
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178605.4 | NP_848720.1 | 5 | 523 | ||
GenBank mRNA | AK002696.1 | - | 5 | 471 | ||
AK003769.1 | - | 5 | 506 | |||
AK013251.1 | - | 5 | 524 | |||
AK088248.1 | - | 5 | 524 | |||
BC012213.1 | - | 5 | 499 | 3964770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
