siRNA Details
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013873.3 | NP_038901.3 | 5 | 704 | ||
GenBank mRNA | AF059257.1 | - | 5 | 580 | ||
AK003034.1 | - | 5 | 565 | |||
AK010293.1 | - | 5 | 560 | |||
AK032195.1 | - | 5 | 590 | |||
AK046368.1 | - | 5 | 583 | |||
AK159034.1 | - | 5 | 596 | |||
BC051132.1 | - | 5 | 577 | 6531493 | ||
BC054757.1 | - | 5 | 704 | 6413084 |

More Information
Other Species Targeted

Gene Ontology Categories:
