siRNA Details
Gene Symbol: Slc39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013901.2 | NP_038929.2 | 4 | 1182 | ||
GenBank mRNA | AK146175.1 | - | 4 | 1174 | ||
AK159290.1 | - | 4 | 1039 | |||
BC005474.1 | - | 4 | 1144 | 3591856 | ||
BC132461.1 | - | 3 | 1119 | 40130738 | ||
BC138459.1 | - | 3 | 1119 | 8861479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
