siRNA Details
Gene Symbol: Slc39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013901.2 | NP_038929.2 | 4 | 1770 | ||
GenBank mRNA | AK159290.1 | - | 4 | 1627 | ||
BC005474.1 | - | 4 | 1732 | 3591856 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
