siRNA Details
Gene Symbol: Hnf4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013920.2 | NP_038948.1 | 2 | 349 | ||
GenBank mRNA | AJ242626.1 | - | 2 | 301 | ||
AK143816.1 | - | 2 | 350 | |||
BC125519.1 | - | 2 | 171 | 40130034 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
