siRNA Details
Gene Symbol: Cxxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004367.4 | NP_001004367.2 | 3 | 1365 | ||
GenBank mRNA | BC065149.1 | - | 4 | 1051 | ||
BC067052.1 | - | 3 | 895 | 30619087 | ||
BC082332.1 | - | 2 | 582 | 30537505 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006233339, XM_008761531, XM_006233341, XM_008761529, XM_008761530, and others.
Gene Ontology Categories:
