siRNA Details
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010568.2 | NP_034698.2 | 21 | 6144 | ||
GenBank mRNA | AK081856.1 | - | 1 | 470 | ||
AK085344.1 | - | 1 | 720 | |||
AK166467.1 | - | 1 | 966 |
