siRNA Details
Gene Symbol: Clpsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034871.2 | NP_001030043.1 | 3 | 327 | ||
GenBank mRNA | AK136682.1 | - | 3 | 329 | ||
BC147116.1 | - | 3 | 403 | 9056160 | ||
BC147117.1 | - | 3 | 403 | 9056121 |
