siRNA Details
Gene Symbol: AI593442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286641.1 | NP_001273570.1 | 2 | 1978 | ||
NM_178906.5 | NP_849237.2 | 2 | 1929 | |||
GenBank mRNA | AK033979.1 | - | 2 | 1923 | ||
AK053489.1 | - | 1 | 65 | |||
CO039428.1 | - | 1 | 83 |
