siRNA Details
Gene Symbol: AI593442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286641.1 | NP_001273570.1 | 2 | 2682 | ||
NM_178906.5 | NP_849237.2 | 2 | 2633 | |||
GenBank mRNA | AK053489.1 | - | 1 | 769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
