siRNA Details
Gene Symbol: Pramel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001319.3 | NP_001001319.2 | 4 | 1510 | ||
GenBank mRNA | AF490340.1 | - | 4 | 1394 | ||
AK145585.1 | - | 4 | 1511 | |||
AK163305.1 | - | 4 | 1369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
