siRNA Details
Gene Symbol: Opn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181753.4 | NP_861418.2 | 5 | 933 | ||
GenBank mRNA | AK039525.1 | - | 1 | 124 | ||
AY288426.1 | - | 5 | 920 | |||
AY318865.1 | - | 5 | 934 | |||
BC125611.1 | - | 5 | 1015 | 40130126 | ||
BC132010.1 | - | 5 | 1015 | 40130287 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
