siRNA Details
Gene Symbol: Opn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181753.4 | NP_861418.2 | 1 | 121 | ||
GenBank mRNA | AY288426.1 | - | 1 | 117 | ||
AY318865.1 | - | 1 | 122 | |||
BC125611.1 | - | 1 | 203 | 40130126 | ||
BC132010.1 | - | 1 | 203 | 40130287 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_181772, AY288431.
Set Membership: |
![]() |
Gene Ontology Categories:
