siRNA Details
Gene Symbol: Hcfc1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181821.1 | NP_861542.1 | 3 | 1157 | ||
GenBank mRNA | AK013438.1 | - | 3 | 1157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
