siRNA Details
Gene Symbol: Nefh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010904.3 | NP_035034.2 | 4 | 1460 | ||
GenBank mRNA | AK122388.1 | - | 4 | 1362 | ||
M35131.1 | - | 5 | 1483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
