siRNA Details
Gene Symbol: Prickle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290337.1 | NP_001277266.1 | 4 | 517 | ||
GenBank mRNA | BX525727.1 | - | 2 | 86 |

More Information
Other Species Targeted

Gene Ontology Categories:
