siRNA Details
Gene Symbol: Lipo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013770.3 | NP_001013792.2 | 6 | 1019 | ||
GenBank mRNA | AK139790.1 | - | 6 | 1020 | ||
AK170332.1 | - | 6 | 953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
