siRNA Details
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK152786.1 | - | 7 | 1434 | ||
BC132314.1 | - | 7 | 1378 | 40130591 | ||
BC132316.1 | - | 7 | 1378 | 40130593 | ||
BC145397.1 | - | 7 | 1372 | 9053955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
