siRNA Details
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 2 | 453 | ||
NM_001301745.1 | NP_001288674.1 | 2 | 453 | |||
GenBank mRNA | AK080485.1 | - | 2 | 402 | ||
AK152278.1 | - | 2 | 322 | |||
AK152683.1 | - | 2 | 321 | |||
AK152786.1 | - | 2 | 330 | |||
AK166217.1 | - | 2 | 453 | |||
BC132314.1 | - | 2 | 274 | 40130591 | ||
BC132316.1 | - | 2 | 274 | 40130593 | ||
BC145397.1 | - | 2 | 274 | 9053955 | ||
M74123.1 | - | 2 | 308 | |||
S62227.1 | - | 2 | 308 |
Gene Symbol: Ifi204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC010546.1 | - | 286 | 4018506 | ||
CT010351.1 | - | 200 | ||||
M31419.1 | - | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
