siRNA Details
Gene Symbol: Tmem240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101506.1 | NP_001094976.1 | 3 | 468 | ||
GenBank mRNA | AK147532.1 | - | 2 | 4527 | ||
BC147820.1 | - | 4 | 329 | 9087537 | ||
BC147826.1 | - | 4 | 329 | 9087543 | ||
BY725301.1 | - | 3 | 400 |
