siRNA Details
Gene Symbol: Gm20809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160137.1 | NP_001153609.1 | 1 | 782 | ||
GenBank mRNA | L04851.1 | - | 1 | 782 |
Gene Symbol: Gm20738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207162.2 | NP_997045.1 | 2 | 1002 | ||
GenBank mRNA | BC049586.1 | - | 2 | 1002 | 6774561 |
Gene Symbol: Gm20747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025241.3 | NP_001020412.1 | 2 | 909 | ||
GenBank mRNA | BC089498.1 | - | 2 | 909 | 6744023 |
Gene Symbol: Gm20816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160144.1 | NP_001153616.1 | 2 | 1011 | ||
GenBank mRNA | BC100413.1 | - | 2 | 1011 | 30919580 |
Gene Symbol: Gm20823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160143.1 | NP_001153615.1 | 2 | 1011 | ||
GenBank mRNA | BC089467.1 | - | 2 | 1011 | 6772630 |
Gene Symbol: Gm20865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160141.1 | NP_001153613.1 | 1 | 731 | ||
GenBank mRNA | L04853.1 | - | 1 | 736 |
Gene Symbol: Gm20867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160142.1 | NP_001153614.1 | 2 | 1011 | ||
GenBank mRNA | BC089466.1 | - | 2 | 1011 | 6742357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
