siRNA Details
Gene Symbol: Gm20865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160141.1 | NP_001153613.1 | 1 | 770 | ||
GenBank mRNA | L04853.1 | - | 1 | 775 |
Gene Symbol: Gm20738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207162.2 | NP_997045.1 | 2 | 1041 | ||
GenBank mRNA | BC049586.1 | - | 2 | 1041 | 6774561 |
Gene Symbol: Gm20747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L04848.1 | - | 820 |
Gene Symbol: Gm20823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160143.1 | NP_001153615.1 | 2 | 1050 | ||
GenBank mRNA | BC089467.1 | - | 2 | 1050 | 6772630 |
Gene Symbol: Gm20826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101623.1 | NP_001095093.1 | 1 | 644 |
Gene Symbol: Gm20867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160142.1 | NP_001153614.1 | 2 | 1050 | ||
GenBank mRNA | BC089466.1 | - | 2 | 1050 | 6742357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
