siRNA Details
Gene Symbol: Olfr128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206816.1 | NP_996552.1 | 1 | 595 | ||
GenBank mRNA | BC139087.1 | - | 1 | 625 |
Gene Symbol: Olfr127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146377.1 | NP_666489.1 | 1 | 595 | ||
GenBank mRNA | BC132433.1 | - | 1 | 621 | 40130710 | |
BC145997.1 | - | 1 | 621 | 40131300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
