siRNA Details
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102409.1 | NP_001095879.1 | 1, 2 | 380 | ||
NM_001102410.1 | NP_001095880.1 | 1, 2 | 380 | |||
NM_201375.2 | NP_958763.1 | 1, 2 | 380 | |||
GenBank mRNA | AY462056.1 | - | 1, 2 | 296 | ||
AY462057.1 | - | 1, 2 | 296 | |||
AY462059.1 | - | 1, 2 | 296 | |||
AY660572.1 | - | 1, 2 | 296 | |||
BC108335.1 | - | 1, 2 | 347 | 5124483 | ||
BC108934.1 | - | 1, 2 | 288 | |||
BC139720.1 | - | 1, 2 | 288 | 40039854 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK128967.1 | - | 211 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
