siRNA Details
Gene Symbol: Plcxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281812.1 | NP_001268741.1 | 7 | 772 | ||
NM_207279.2 | NP_997162.2 | 8 | 937 | |||
GenBank mRNA | AK039459.1 | - | 6 | 677 | ||
AK149435.1 | - | 7 | 773 | |||
BC034116.1 | - | 5 | 630 | |||
BC039627.1 | - | 8 | 937 | 5038680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
