siRNA Details
Gene Symbol: Mrgprx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207540.3 | NP_997423.1 | 2 | 1011 | ||
GenBank mRNA | AY152435.1 | - | 2 | 1017 | ||
BC150669.1 | - | 2 | 961 | 9087580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
