siRNA Details
Gene Symbol: Olfr373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146539.2 | NP_666750.2 | 1 | 271 | ||
GenBank mRNA | BC106979.2 | - | 1 | 269 | ||
BC106980.1 | - | 1 | 268 | 40042607 | ||
BC119444.1 | - | 1 | 271 | |||
BC119445.2 | - | 1 | 271 | 40042612 |
Gene Symbol: Olfr372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207555.2 | NP_997438.1 | 1 | 293 | ||
GenBank mRNA | BC150671.1 | - | 1 | 293 | 9087582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
