siRNA Details
Gene Symbol: Krt83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003668.2 | NP_001003668.2 | 4 | 743 | ||
GenBank mRNA | BC139321.1 | - | 4 | 743 | 8862343 | |
BC139322.1 | - | 4 | 743 | 8862344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
