siRNA Details
Gene Symbol: Ndufa4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098789.1 | NP_001092259.1 | 4 | 297 | ||
GenBank mRNA | AK131609.1 | - | 4 | 269 | ||
AK131662.1 | - | 4 | 266 | |||
BC064011.1 | - | 4 | 258 | |||
BC099496.1 | - | 4 | 258 | 30847512 | ||
BY336656.1 | - | 4 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
