siRNA Details
Gene Symbol: BC048507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001185.3 | NP_001001185.1 | 1 | 453 | ||
GenBank mRNA | AK133011.1 | - | 1 | 454 | ||
BC048507.1 | - | 1 | 442 | 6774067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
