siRNA Details
Gene Symbol: Zfp729b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163246.2 | NP_001156718.1 | 3 | 287 | ||
GenBank mRNA | AK137425.1 | - | 3 | 288 | ||
BB629152.1 | - | 3 | 288 | |||
BC047145.1 | - | 3 | 243 | 6467898 |
Gene Symbol: Zfp65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145622.2 | NP_663597.1 | 3 | 265 | ||
GenBank mRNA | AK041786.1 | - | 3 | 212 | ||
BB621429.1 | - | 3 | 266 | |||
BC016248.1 | - | 2 | 174 | 4527362 | ||
U29507.1 | - | 2 | 71 |
Gene Symbol: Zfp738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001187.3 | NP_001001187.2 | 3 | 262 | ||
GenBank mRNA | AK076179.1 | - | 3 | 266 | ||
BC082605.1 | - | 3 | 217 | |||
BY752525.1 | - | 3 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
