siRNA Details
Gene Symbol: Gm5415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164286.1 | NP_001157758.1 | 1 | 2090 | ||
GenBank mRNA | AK029494.1 | - | 1 | 2092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
