siRNA Details
Gene Symbol: Tmem95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195710.1 | NP_001182639.1 | 6 | 445 | ||
GenBank mRNA | BF018485.1 | - | 6 | 392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
