siRNA Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033488.2 | NP_001028660.2 | 9 | 933 | ||
GenBank mRNA | AK133061.1 | - | 9 | 948 | ||
AK156138.1 | - | 10 | 1085 | |||
AK157892.1 | - | 8 | 1760 | |||
AK162620.1 | - | 9 | 932 | |||
AK171660.1 | - | 6 | 596 | |||
AK172423.1 | - | 9 | 934 | |||
BC145584.1 | - | 9 | 946 |
