siRNA Details
Gene Symbol: Gm21319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270722.1 | NP_001257651.1 | 2 | 931 | ||
GenBank mRNA | AK139496.1 | - | 2 | 933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
