siRNA Details
Gene Symbol: Snrpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026943.1 | NP_081219.1 | 3 | 299 | ||
GenBank mRNA | AK007389.1 | - | 3 | 278 | ||
AK045968.1 | - | 3 | 274 | |||
AK088105.1 | - | 3 | 278 | |||
BC043014.1 | - | 3 | 299 | 5683364 | ||
BC051208.1 | - | 2 | 278 | 1510565 | ||
BC116858.1 | - | 3 | 282 | 40126177 | ||
BC116860.1 | - | 3 | 282 | 40126179 |

More Information
Other Species Targeted

Gene Ontology Categories:
