siRNA Details
Gene Symbol: Defb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037751.3 | NP_001032840.1 | 2 | 345 | ||
GenBank mRNA | AK136677.1 | - | 2 | 346 | ||
BY721060.1 | - | 2 | 346 | |||
DQ012044.1 | - | 2 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
