siRNA Details
Gene Symbol: Defb45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037752.2 | NP_001032841.1 | 2 | 325 | ||
GenBank mRNA | AK005919.1 | - | 2 | 326 | ||
BC132226.1 | - | 2 | 265 | 40130503 | ||
BC132228.1 | - | 2 | 265 | 40130505 | ||
DQ012038.1 | - | 2 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
