siRNA Details
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291865.1 | NP_001278794.1 | 5 | 684 | ||
NM_009121.4 | NP_033147.1 | 4 | 574 | |||
GenBank mRNA | AK002531.1 | - | 4 | 475 | ||
AK131657.1 | - | 1 | 861 | |||
AK154445.1 | - | 4 | 470 | |||
BC058696.1 | - | 4 | 534 | 6820169 | ||
BC108334.1 | - | 4 | 885 | |||
L10244.1 | - | 4 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
