siRNA Details
Gene Symbol: Dppa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025274.3 | NP_079550.2 | 2 | 387 | ||
GenBank mRNA | AF490349.1 | - | 2 | 343 | ||
AK010519.1 | - | 2 | 344 | |||
AK010584.1 | - | 2 | 343 | |||
AK145761.1 | - | 2 | 346 | |||
AV457404.1 | - | 2 | 389 | |||
BC092353.1 | - | 2 | 332 | 30730511 | ||
BC092354.1 | - | 2 | 340 | 30732471 | ||
BC095979.1 | - | 2 | 332 | 30732937 | ||
X57708.1 | - | 2 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
