siRNA Details
Gene Symbol: Mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290810.1 | NP_001277739.1 | 4 | 527 | ||
NM_023536.3 | NP_076025.1 | 4 | 527 | |||
GenBank mRNA | AK011387.1 | - | 4 | 249 | ||
AK171445.1 | - | 2 | 1509 | |||
AK192789.1 | - | 1 | 58 | |||
BC005734.1 | - | 4 | 249 | 3598883 | ||
BC096556.1 | - | 4 | 271 | |||
BC138221.1 | - | 4 | 326 | 8861240 | ||
BC145205.1 | - | 4 | 326 | 9053756 | ||
BC145209.1 | - | 4 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
