siRNA Details
Gene Symbol: Rhox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004193.2 | NP_001004193.1 | 1, 2 | 219 | ||
GenBank mRNA | AY329370.1 | - | 1, 2 | 219 | ||
BC139098.1 | - | 1, 2 | 202 | 8862120 | ||
DQ058645.1 | - | 2, 3 | 228 |
