siRNA Details
Gene Symbol: Taar7d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010838.1 | NP_001010838.1 | 1 | 709 |
Gene Symbol: Taar7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010829.1 | NP_001010829.1 | 1 | 709 |
Gene Symbol: Taar7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010827.1 | NP_001010827.1 | 1 | 709 |
Gene Symbol: Taar7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010835.1 | NP_001010835.1 | 1 | 709 | ||
GenBank mRNA | BC139110.1 | - | 1 | 775 | 8862132 | |
BC145816.1 | - | 1 | 775 | 40131119 |
Gene Symbol: Taar7f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010839.1 | NP_001010839.1 | 1 | 709 | ||
GenBank mRNA | BC139175.1 | - | 1 | 737 | 8862197 | |
BC139176.1 | - | 1 | 737 | 8862198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
