siRNA Details
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011646.5 | NP_035776.1 | 2, 3 | 213 | ||
GenBank mRNA | AK003064.1 | - | 2, 3 | 217 | ||
AK008660.1 | - | 2, 3 | 214 | |||
AK008667.1 | - | 2, 3 | 214 | |||
BC050120.1 | - | 2, 3 | 194 | |||
BC061135.1 | - | 2, 3 | 213 | 30306436 |
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038996.2 | NP_001034085.1 | 2, 3 | 287 | ||
GenBank mRNA | AK133516.1 | - | 2, 3 | 214 | ||
BC138698.1 | - | 2, 3 | 287 | 8861720 | ||
BC146017.1 | - | 2, 3 | 287 | 40131320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
