siRNA Details
Gene Symbol: Scgb2b20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009952.2 | NP_001009952.2 | 3 | 290 | ||
GenBank mRNA | AF272844.1 | - | 3 | 294 | ||
CK616187.1 | - | 3 | 291 |
Gene Symbol: Scgb2b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198871.1 | NP_001185800.1 | 3 | 287 | ||
GenBank mRNA | CK615801.1 | - | 3 | 288 | ||
CK616838.1 | - | 3 | 245 | |||
CK616903.1 | - | 3 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
