siRNA Details
Gene Symbol: Rnase13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011687.2 | NP_001011687.1 | 1 | 268 | ||
GenBank mRNA | BC132614.1 | - | 1 | 268 | 40130891 | |
BC132616.1 | - | 1 | 268 | 40130893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
