siRNA Details
Gene Symbol: Klri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012520.2 | NP_001012538.2 | 6 | 690 | ||
GenBank mRNA | AY324874.1 | - | 6 | 690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
