siRNA Details
Gene Symbol: Mrps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025305.2 | NP_079581.1 | 5 | 601 | ||
GenBank mRNA | AK009251.1 | - | 5 | 566 | ||
AK012225.1 | - | 5 | 601 | |||
AK131937.1 | - | 5 | 612 | |||
AK162024.1 | - | 5 | 580 | |||
BC049636.1 | - | 5 | 601 | 6532964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
