siRNA Details
Gene Symbol: Dkkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015789.3 | NP_056604.2 | 5 | 854 | ||
GenBank mRNA | AF177399.1 | - | 5 | 759 | ||
AF274312.1 | - | 5 | 813 | |||
AK131694.1 | - | 5 | 799 | |||
BC002215.1 | - | 5 | 832 | 3490482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
